siRNA Details
| Gene Symbol: Otud4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081164.1 | NP_001074633.1 | 21 | 3153 | ||
| NM_001256033.1 | NP_001242962.1 | 21 | 3156 | |||
| GenBank mRNA | AK015275.1 | - | 1 | 141 | ||
| AK122429.1 | - | 16 | 2447 | |||
| BC138373.1 | - | 21 | 3128 | 8861393 | ||
| BC145259.1 | - | 21 | 3125 | 9053812 | ||

Otud4
Genomic Map