siRNA Details
| Gene Symbol: Rftn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028713.1 | NP_082989.1 | 4 | 990 | ||
| GenBank mRNA | AK012229.2 | - | 2 | 688 | ||
| AK014318.1 | - | 4 | 742 | |||
| BC026871.1 | - | 4 | 554 | 3966594 | ||
| BC038341.1 | - | 4 | 990 | 4218473 | ||

Rftn2
Genomic Map