siRNA Details
| Gene Symbol: Rftn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028713.1 | NP_082989.1 | 3 | 896 | ||
| GenBank mRNA | AK014318.1 | - | 3 | 648 | ||
| BC026871.1 | - | 3 | 460 | 3966594 | ||
| BC038341.1 | - | 3 | 896 | 4218473 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rftn2| Gene Symbol: Rftn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028713.1 | NP_082989.1 | 3 | 896 | ||
| GenBank mRNA | AK014318.1 | - | 3 | 648 | ||
| BC026871.1 | - | 3 | 460 | 3966594 | ||
| BC038341.1 | - | 3 | 896 | 4218473 | ||