siRNA Details
| Gene Symbol: Traf3ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028718.2 | NP_082994.1 | 7 | 1108 | ||
| GenBank mRNA | AK014457.1 | - | 4 | 589 | ||
| AY613437.1 | - | 7 | 1111 | |||
| BC117868.2 | - | 7 | 1082 | 40095476 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Traf3ip1| Gene Symbol: Traf3ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028718.2 | NP_082994.1 | 7 | 1108 | ||
| GenBank mRNA | AK014457.1 | - | 4 | 589 | ||
| AY613437.1 | - | 7 | 1111 | |||
| BC117868.2 | - | 7 | 1082 | 40095476 | ||