siRNA Details
Gene Symbol: Cpne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028719.1 | NP_082995.1 | 2 | 356 | ||
GenBank mRNA | AK014396.1 | - | 2 | 358 | ||
AK016705.1 | - | 5 | 891 | |||
AK043692.1 | - | 2 | 369 | |||
BC043087.1 | - | 2 | 356 | 6417611 | ||
BC063081.1 | - | 2 | 356 | 6417611 |
