siRNA Details
| Gene Symbol: Ddias | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080995.1 | NP_001074464.1 | 4 | 347 | ||
| GenBank mRNA | AK014608.2 | - | 3 | 267 | ||
| AK154915.1 | - | 3 | 250 | |||
| BC066148.1 | - | 3 | 241 | 30086532 | ||
| DQ400346.1 | - | 3 | 281 | |||

Ddias
Genomic Map