siRNA Details
| Gene Symbol: 4921501E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009544.2 | NP_001009544.1 | 1 | 1462 | ||
| GenBank mRNA | AK077044.1 | - | 1 | 1430 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4921501E09Rik| Gene Symbol: 4921501E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009544.2 | NP_001009544.1 | 1 | 1462 | ||
| GenBank mRNA | AK077044.1 | - | 1 | 1430 | ||