siRNA Details
| Gene Symbol: Ttc21a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028735.3 | NP_083011.2 | 24 | 3332 | ||
| GenBank mRNA | AK015017.1 | - | 24 | 3292 | ||
| AK029952.1 | - | 24 | 3336 | |||
| AK133304.1 | - | 24 | 3304 | |||
| BC133699.1 | - | 24 | 3290 | 40098338 | ||
| DQ011665.1 | - | 24 | 3336 | |||

Ttc21a
Genomic Map