siRNA Details
| Gene Symbol: Plce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019588.2 | NP_062534.2 | 21 | 5002 | ||
| GenBank mRNA | AB076247.1 | - | 21 | 5002 | ||
| AK033248.1 | - | 5 | 3616 | |||
| AK122521.1 | - | 15 | 2188 | |||
| BC138349.1 | - | 21 | 4990 | 8861369 | ||
| BC138350.1 | - | 21 | 4990 | 8861370 | ||
| BC145246.1 | - | 22 | 5032 | 9053798 | ||

Plce1
Genomic Map