siRNA Details
| Gene Symbol: Ifltd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028742.2 | NP_083018.1 | 5 | 556 | ||
| GenBank mRNA | AK016641.1 | - | 5 | 556 | ||
| AK052206.1 | - | 3 | 635 | |||
| AY753326.1 | - | 3 | 391 | |||
| BC053420.1 | - | 4 | 518 | 6705208 | ||

Ifltd1
Genomic Map