siRNA Details
| Gene Symbol: Hvcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042489.2 | NP_001035954.1 | 6 | 835 | ||
| NM_028752.3 | NP_083028.1 | 5 | 697 | |||
| GenBank mRNA | AK002854.1 | - | 5 | 698 | ||
| AK154880.1 | - | 6 | 727 | |||
| AK170422.1 | - | 6 | 696 | |||
| BC021548.1 | - | 6 | 666 | 4913027 | ||
| BY703352.1 | - | 5 | 691 | |||
| CF734112.1 | - | 6 | 670 | |||

Hvcn1
Genomic Map