siRNA Details
| Gene Symbol: Abcb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023732.2 | NP_076221.1 | 1 | 239 | ||
| GenBank mRNA | AF045562.1 | - | 1 | 92 | ||
| AK004605.1 | - | 1 | 319 | |||
| AK040323.1 | - | 1 | 243 | |||
| AK150853.1 | - | 1 | 327 | |||
| AK151165.1 | - | 1 | 349 | |||
| AK152584.1 | - | 1 | 327 | |||
| AK164003.1 | - | 1 | 243 | |||
| AK168642.1 | - | 1 | 340 | |||
| BC006634.1 | - | 1 | 239 | 3710552 | ||

Abcb6
Genomic Map