siRNA Details
| Gene Symbol: Gga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028758.2 | NP_083034.1 | 5 | 572 | ||
| GenBank mRNA | AK004632.1 | - | 5 | 544 | ||
| AK034612.1 | - | 2 | 215 | |||
| AK129280.1 | - | 5 | 531 | |||
| AK143691.1 | - | 5 | 560 | |||
| AK148205.1 | - | 555 | ||||
| AK149798.1 | - | 5 | 562 | |||
| AK154904.1 | - | 5 | 547 | |||
| AK159953.1 | - | 5 | 562 | |||
| AK196219.1 | - | 1 | 26 | |||
| BC062933.1 | - | 5 | 541 | 30356060 | ||

Gga2
Genomic Map