siRNA Details
| Gene Symbol: Cep55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164362.1 | NP_001157834.1 | 9 | 1561 | ||
| NM_028293.1 | NP_082569.1 | 8 | 1185 | |||
| NM_028760.2 | NP_083036.2 | 9 | 1537 | |||
| GenBank mRNA | AK004655.1 | - | 9 | 1563 | ||
| AK028216.1 | - | 8 | 1171 | |||
| BC026966.1 | - | 6 | 893 | |||
| BC031396.1 | - | 9 | 1490 | 4950530 | ||

Cep55
Genomic Map