siRNA Details
| Gene Symbol: Parn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028761.3 | NP_083037.1 | 7 | 649 | ||
| GenBank mRNA | AK004572.1 | - | 7 | 650 | ||
| AK041044.1 | - | 7 | 618 | |||
| AK045181.1 | - | 7 | 635 | |||
| AK049829.1 | - | 5 | 1725 | |||
| AK160611.1 | - | 7 | 618 | |||
| BC021899.1 | - | 7 | 617 | 4015784 | ||

Parn
Genomic Map