siRNA Details
| Gene Symbol: Zfp263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148924.3 | NP_683726.2 | 6 | 1929 | ||
| GenBank mRNA | AF499776.1 | - | 6 | 1897 | ||
| AK004765.2 | - | 6 | 1926 | |||
| AK159455.1 | - | 6 | 1917 | |||
| BC011505.1 | - | 7 | 2455 | |||
| BC029993.1 | - | 2 | 1129 | |||
| BC056222.1 | - | 6 | 1750 | 30009326 | ||
| BC060162.1 | - | 5 | 2862 | |||

Zfp263
Genomic Map