siRNA Details
| Gene Symbol: Dmgdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028772.3 | NP_083048.1 | 16 | 2562 | ||
| GenBank mRNA | AK004755.1 | - | 16 | 2563 | ||
| BC024126.1 | - | 12 | 1845 | |||
| BC089599.1 | - | 16 | 2565 | 30118948 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dmgdh| Gene Symbol: Dmgdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028772.3 | NP_083048.1 | 16 | 2562 | ||
| GenBank mRNA | AK004755.1 | - | 16 | 2563 | ||
| BC024126.1 | - | 12 | 1845 | |||
| BC089599.1 | - | 16 | 2565 | 30118948 | ||