siRNA Details
| Gene Symbol: Smg8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024262.1 | NP_077224.1 | 2 | 1902 | ||
| GenBank mRNA | AK004714.1 | - | 2 | 1895 | ||
| AK031106.1 | - | 2 | 1876 | |||
| AK035089.1 | - | 2 | 869 | |||
| BC002247.1 | - | 2 | 507 | |||
| BC020005.1 | - | 2 | 1902 | 3981459 | ||

Smg8
Genomic Map