siRNA Details
| Gene Symbol: Robo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF536772.1 | - | 7 | 1173 | ||
| AK004723.2 | - | 7 | 1170 | |||
| AK087355.1 | - | 7 | 1167 | |||
| AK172104.1 | - | 6 | 1411 | |||
| BC071193.1 | - | 6 | 863 | 5060473 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Robo4| Gene Symbol: Robo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF536772.1 | - | 7 | 1173 | ||
| AK004723.2 | - | 7 | 1170 | |||
| AK087355.1 | - | 7 | 1167 | |||
| AK172104.1 | - | 6 | 1411 | |||
| BC071193.1 | - | 6 | 863 | 5060473 | ||