siRNA Details
Gene Symbol: Stra6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004855.1 | - | 5, 6 | 562 | ||
AK045033.1 | - | 6, 7 | 990 | |||
AK050152.1 | - | 5, 6 | 573 | |||
AK050348.1 | - | 5, 6 | 569 | |||
BC111889.1 | - | 5, 6 | 499 | 40060643 | ||
BC112408.1 | - | 5, 6 | 499 | 40060646 |
