siRNA Details
Gene Symbol: Stra6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004855.1 | - | 4 | 375 | ||
AK045033.1 | - | 5 | 803 | |||
AK050152.1 | - | 4 | 386 | |||
AK050348.1 | - | 4 | 382 | |||
BC111889.1 | - | 4 | 312 | 40060643 | ||
BC112408.1 | - | 4 | 312 | 40060646 |
