siRNA Details
| Gene Symbol: Nfx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290448.1 | NP_001277377.1 | 9 | 1927 | ||
| NM_001290449.1 | NP_001277378.1 | 9 | 1927 | |||
| NM_023739.3 | NP_076228.2 | 9 | 1890 | |||
| GenBank mRNA | AF223576.1 | - | 9 | 1854 | ||
| AK005038.2 | - | 9 | 1862 | |||
| AK033850.1 | - | 9 | 1821 | |||
| AK050749.1 | - | 9 | 1864 | |||
| AK144647.1 | - | 9 | 1876 | |||
| AK146108.1 | - | 9 | 1844 | |||
| AK155387.1 | - | 9 | 1842 | |||
| AK160013.1 | - | 9 | 1860 | |||
| BC053709.1 | - | 9 | 1825 | 30052746 | ||

Nfx1
Genomic Map