siRNA Details
| Gene Symbol: Stk40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145827.1 | NP_001139299.1 | 10 | 1344 | ||
| NM_028800.3 | NP_083076.3 | 8 | 990 | |||
| GenBank mRNA | AB102946.1 | - | 8 | 821 | ||
| AK009154.1 | - | 3 | 208 | |||
| AK050808.1 | - | 4 | 293 | |||
| AK154780.1 | - | 10 | 1335 | |||
| AK197351.1 | - | 1 | 42 | |||
| AY336057.1 | - | 8 | 975 | |||
| BC046981.1 | - | 8 | 949 | 6478265 | ||

Stk40
Genomic Map