siRNA Details
| Gene Symbol: Gbe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028803.4 | NP_083079.1 | 2 | 392 | ||
| GenBank mRNA | AK009815.1 | - | 2 | 208 | ||
| AK013167.1 | - | 2 | 201 | |||
| AK050365.1 | - | 2 | 279 | |||
| AK050423.1 | - | 2 | 278 | |||
| AK206164.1 | - | 2 | 63 | |||
| BC017541.1 | - | 2 | 253 | 4526961 | ||

Gbe1
Genomic Map