siRNA Details
| Gene Symbol: Gbe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028803.4 | NP_083079.1 | 3, 4 | 629 | ||
| GenBank mRNA | AK009815.1 | - | 3, 4 | 445 | ||
| AK050365.1 | - | 3, 4 | 516 | |||
| AK050423.1 | - | 3, 4 | 515 | |||
| AK206164.1 | - | 3, 4 | 300 | |||
| BC017541.1 | - | 3, 4 | 490 | 4526961 | ||

Gbe1
Genomic Map