siRNA Details
| Gene Symbol: Ttc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152817.4 | NP_690030.3 | 19 | 2543 | ||
| GenBank mRNA | AI118251.1 | - | 2 | 293 | ||
| AK012131.1 | - | 19 | 2543 | |||
| AK030942.1 | - | 19 | 2544 | |||
| AK049324.1 | - | 19 | 2544 | |||
| BC021912.1 | - | 18 | 2278 | 4503335 | ||

Ttc27
Genomic Map