siRNA Details
| Gene Symbol: Ttc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152817.4 | NP_690030.3 | 9, 10 | 1285 | ||
| GenBank mRNA | AK012131.1 | - | 9, 10 | 1285 | ||
| AK030942.1 | - | 9, 10 | 1286 | |||
| AK049324.1 | - | 9, 10 | 1286 | |||
| AK168138.1 | - | 9, 10 | 1276 | |||
| BC021912.1 | - | 8, 9 | 1020 | 4503335 | ||

Ttc27
Genomic Map