siRNA Details
| Gene Symbol: 2810403A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028814.3 | NP_083090.2 | 7 | 852 | ||
| GenBank mRNA | AK012969.1 | - | 7 | 829 | ||
| AK083018.1 | - | 7 | 813 | |||
| AK087126.1 | - | 1 | 520 | |||
| AK170490.1 | - | 7 | 847 | |||
| AK173057.1 | - | 7 | 816 | |||
| AK193095.1 | - | 1 | 66 | |||
| BC006621.1 | - | 7 | 827 | 3158104 | ||

2810403A07Rik
Genomic Map