siRNA Details
| Gene Symbol: 2810403A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028814.3 | NP_083090.2 | 5, 6 | 590 | ||
| GenBank mRNA | AK012969.1 | - | 5, 6 | 567 | ||
| AK083018.1 | - | 5, 6 | 551 | |||
| AK170490.1 | - | 5, 6 | 585 | |||
| AK173057.1 | - | 5, 6 | 554 | |||
| BC006621.1 | - | 5, 6 | 565 | 3158104 | ||
| CX562176.1 | - | 5, 6 | 556 | |||

2810403A07Rik
Genomic Map