siRNA Details
Gene Symbol: 1700017B05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028820.2 | NP_083096.1 | 2 | 1815 | ||
GenBank mRNA | AK029230.1 | - | 1819 | |||
AK031730.1 | - | 2 | 1818 | |||
AK044396.1 | - | 2 | 1820 | |||
AK080676.1 | - | 2 | 1828 | |||
AK145074.1 | - | 2 | 1792 | |||
AK155015.1 | - | 2 | 1792 | |||
AK155487.1 | - | 2 | 1816 | |||
AK165963.1 | - | 2 | 1816 | |||
AK169617.1 | - | 2 | 1818 | |||
AK190772.1 | - | 1 | 24 | |||
BC118527.1 | - | 2 | 1669 | 40098810 | ||
BC150963.1 | - | 2 | 1660 | 9087874 | ||
BC150965.1 | - | 2 | 1660 | 9087876 |
