siRNA Details
| Gene Symbol: Cyld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128170.2 | NP_001121642.1 | 6 | 1173 | ||
| NM_001128171.2 | NP_001121643.1 | 6 | 1298 | |||
| NM_001276279.1 | NP_001263208.1 | 6 | 1295 | |||
| NM_173369.3 | NP_775545.1 | 5 | 1164 | |||
| GenBank mRNA | AK039054.1 | - | 7 | 1302 | ||
| AK122389.1 | - | 6 | 1279 | |||
| AK133479.1 | - | 5 | 1128 | |||
| AK147204.1 | - | 6 | 1142 | |||
| AK147426.1 | - | 6 | 1137 | |||
| AK167424.1 | - | 6 | 1259 | |||
| BC042438.1 | - | 5 | 1117 | 3983771 | ||

Cyld
Genomic Map