siRNA Details
| Gene Symbol: Tspan17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028841.3 | NP_083117.2 | 8 | 949 | ||
| GenBank mRNA | AI019499.1 | - | 5 | 419 | ||
| AK008761.1 | - | 8 | 894 | |||
| AK166272.1 | - | 8 | 952 | |||
| AK171691.1 | - | 8 | 898 | |||
| BC010346.1 | - | 8 | 894 | 2811935 | ||
| CD740983.1 | - | 2 | 236 | |||

Tspan17
Genomic Map