siRNA Details
| Gene Symbol: Rnf138rt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028842.3 | NP_083118.1 | 1 | 735 | ||
| GenBank mRNA | AI853461.1 | - | 1 | 472 | ||
| AK006700.1 | - | 1 | 734 | |||
| BC125311.1 | - | 1 | 663 | |||
| BC125313.1 | - | 1 | 663 | |||

Rnf138rt1
Genomic Map