siRNA Details
| Gene Symbol: Spem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028855.1 | NP_083131.1 | 3 | 867 | ||
| GenBank mRNA | AK007080.1 | - | 3 | 883 | ||
| BC048760.1 | - | 3 | 867 | |||
| BC100505.1 | - | 3 | 859 | 30917608 | ||
| BC144838.1 | - | 1 | 1231 | |||
| EF120626.1 | - | 3 | 867 | |||

Spem1
Genomic Map