siRNA Details
| Gene Symbol: Mtmr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028860.2 | NP_083136.2 | 5 | 546 | ||
| GenBank mRNA | BC032166.1 | - | 5 | 529 | 5352919 | |
| BC081544.1 | - | 5 | 530 | 3672099 | ||
| CJ048010.1 | - | 5 | 547 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtmr3| Gene Symbol: Mtmr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028860.2 | NP_083136.2 | 5 | 546 | ||
| GenBank mRNA | BC032166.1 | - | 5 | 529 | 5352919 | |
| BC081544.1 | - | 5 | 530 | 3672099 | ||
| CJ048010.1 | - | 5 | 547 | |||