siRNA Details
| Gene Symbol: Palm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028877.1 | NP_083153.1 | 7 | 1162 | ||
| GenBank mRNA | AK014493.1 | - | 6 | 2207 | ||
| BC113196.1 | - | 7 | 1165 | |||
| BC113802.1 | - | 7 | 1160 | |||
| EF208034.1 | - | 7 | 1162 | |||

Palm3
Genomic Map