siRNA Details
| Gene Symbol: Lrrtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028880.3 | NP_083156.2 | 2 | 2071 | ||
| GenBank mRNA | AK014545.1 | - | 2 | 2073 | ||
| AK053345.1 | - | 2 | 2061 | |||
| AK053364.1 | - | 2 | 1695 | |||
| AK080836.1 | - | 2 | 2102 | |||
| AK136857.1 | - | 1 | 1570 | |||
| AY182025.1 | - | 1 | 1506 | |||
| BC027803.1 | - | 2 | 1953 | 5321979 | ||
| BQ176603.1 | - | 1 | 462 | |||

Lrrtm1
Genomic Map