siRNA Details
| Gene Symbol: Smchd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028887.3 | NP_083163.3 | 35, 36 | 4620 | ||
| GenBank mRNA | AK016419.1 | - | 10, 11 | 1097 | ||
| AK030247.1 | - | 3, 4 | 201 | |||
| AK129181.1 | - | 13, 14 | 1617 | |||
| BC058205.1 | - | 1, 2 | 80 | |||
| BC058618.1 | - | 11, 12 | 1272 | |||
| BC062946.1 | - | 21, 22 | 2465 | 30357469 | ||
| BC076627.1 | - | 28, 29 | 3583 | |||

Smchd1
Genomic Map