siRNA Details
Gene Symbol: 4931414P19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028890.2 | NP_083166.1 | 3 | 969 | ||
GenBank mRNA | AK016457.1 | - | 2 | 442 | ||
AK163117.1 | - | 3 | 978 | |||
BC029624.1 | - | 3 | 969 | 3992239 | ||
BC034212.1 | - | 3 | 969 |
