siRNA Details
Gene Symbol: 4931414P19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028890.2 | NP_083166.1 | 2 | 521 | ||
GenBank mRNA | AK163117.1 | - | 2 | 530 | ||
BC029624.1 | - | 2 | 521 | 3992239 | ||
BC034212.1 | - | 2 | 521 |
