siRNA Details
| Gene Symbol: Cep57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026665.4 | NP_080941.3 | 9 | 1264 | ||
| GenBank mRNA | AK014862.1 | - | 6 | 897 | ||
| AK014873.1 | - | 9 | 1211 | |||
| AK014982.1 | - | 7 | 722 | |||
| AK016484.1 | - | 9 | 1048 | |||
| AK019414.1 | - | 9 | 1144 | |||
| AK028408.1 | - | 8 | 1036 | |||
| AY225093.1 | - | 9 | 1057 | |||
| AY251192.1 | - | 9 | 1209 | |||
| BC025588.1 | - | 8 | 1044 | |||
| BC050785.1 | - | 7 | 775 | 6703205 | ||
| BC056968.1 | - | 9 | 1508 | |||

Cep57
Genomic Map