siRNA Details
| Gene Symbol: Map10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028908.3 | NP_083184.1 | 1 | 2686 | ||
| GenBank mRNA | AK016632.1 | - | 1 | 2642 | ||
| AK042358.1 | - | 1 | 2659 | |||
| AK080043.1 | - | 1 | 2640 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map10| Gene Symbol: Map10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028908.3 | NP_083184.1 | 1 | 2686 | ||
| GenBank mRNA | AK016632.1 | - | 1 | 2642 | ||
| AK042358.1 | - | 1 | 2659 | |||
| AK080043.1 | - | 1 | 2640 | |||