siRNA Details
| Gene Symbol: Zfp819 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028913.3 | NP_083189.2 | 5 | 532 | ||
| GenBank mRNA | AK015213.1 | - | 4 | 339 | ||
| AK016669.1 | - | 5 | 477 | |||
| BC050101.1 | - | 4 | 414 | 30007787 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp819| Gene Symbol: Zfp819 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028913.3 | NP_083189.2 | 5 | 532 | ||
| GenBank mRNA | AK015213.1 | - | 4 | 339 | ||
| AK016669.1 | - | 5 | 477 | |||
| BC050101.1 | - | 4 | 414 | 30007787 | ||