siRNA Details
Gene Symbol: Efhc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028916.4 | NP_083192.2 | 5 | 908 | ||
GenBank mRNA | AK016714.1 | - | 5 | 867 | ||
AK029501.1 | - | 5 | 874 | |||
BC120659.1 | - | 5 | 843 | 40129582 | ||
BC125625.1 | - | 5 | 843 | 40130140 |
