siRNA Details
| Gene Symbol: Ttc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028918.2 | NP_083194.2 | 7 | 1072 | ||
| GenBank mRNA | AK016657.1 | - | 7 | 1003 | ||
| AK161541.1 | - | 7 | 1011 | |||
| AK162855.1 | - | 7 | 1012 | |||
| BC050805.1 | - | 7 | 1096 | 6771545 | ||

Ttc25
Genomic Map