siRNA Details
| Gene Symbol: 4932442L08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016569.1 | - | 6 | 1662 | ||
| BC115707.1 | - | 4 | 1300 | 40058141 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4932442L08Rik| Gene Symbol: 4932442L08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016569.1 | - | 6 | 1662 | ||
| BC115707.1 | - | 4 | 1300 | 40058141 | ||