siRNA Details
| Gene Symbol: 4933427D14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028963.2 | NP_083239.1 | 16 | 2624 | ||
| GenBank mRNA | AK032379.1 | - | 16 | 2502 | ||
| AK036794.1 | - | 10 | 1235 | |||
| AK041396.1 | - | 17 | 2818 | |||
| AK173018.1 | - | 16 | 2493 | |||
| BC043106.1 | - | 16 | 2624 | 6406803 | ||
| BC059269.1 | - | 17 | 2777 | |||

4933427D14Rik
Genomic Map