siRNA Details
| Gene Symbol: Snx29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290148.1 | NP_001277077.1 | 13 | 1282 | ||
| NM_028964.4 | NP_083240.2 | 20 | 2331 | |||
| GenBank mRNA | AF399755.1 | - | 13 | 1237 | ||
| AK017102.1 | - | 13 | 1283 | |||

Snx29
Genomic Map