siRNA Details
| Gene Symbol: Snx29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290148.1 | NP_001277077.1 | 14 | 1401 | ||
| NM_028964.4 | NP_083240.2 | 21 | 2450 | |||
| GenBank mRNA | AF399755.1 | - | 14 | 1356 | ||
| AK017102.1 | - | 14 | 1402 | |||
| CR521963.1 | - | 1 | 224 | |||

Snx29
Genomic Map