siRNA Details
| Gene Symbol: Snx29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290148.1 | NP_001277077.1 | 4, 5 | 418 | ||
| NM_028964.4 | NP_083240.2 | 11, 12 | 1467 | |||
| GenBank mRNA | AF399755.1 | - | 4, 5 | 373 | ||
| AK017102.1 | - | 4, 5 | 419 | |||
| AK162238.1 | - | 3, 4 | 296 | |||
| BB623950.1 | - | 3, 4 | 296 | |||
| BC034114.1 | - | 1, 2 | 67 | |||

Snx29
Genomic Map